Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, κ Light Chain Mouse anti-Human, BB515, Clone: G20-193, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565232

Catalog No. BDB565232


Add to cart

Description

Description

The G20-193 monoclonal antibody specifically binds to human immunoglobulin light chain, kappa ( κ ). It does not react with human immunoglobulin λ light chains or human immunoglobulin heavy chains.

Specifications

Specifications

Ig, κ Light Chain
G20-193
Ig, kappa Light Chain; IGKC; HCAK1; Km; Ig kappa chain C region
Affinity purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide
Murine
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit