Learn More
Kappa, Ig light chain Mouse anti-Human, FITC, Clone: TB28-2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 643773
Catalog No. BDB643773
Add to cart
Description
Description
Anti-Kappa is specific for kappa light chains of human immunoglobulins. Immunoglobulins bearing kappa light chains are present on approximately 60% of normal B lymphocytes and on Igκ+ neoplastic cells. In serum, Anti-Kappa reacts with immunoglobulins bearing kappa light chains as well as free kappa light chains.
Flow Cytometry
Specifications
Specifications
Kappa, Ig light chain
TB28-2
FITC
Conjugated
Human IgG, κ myeloma protein
50 Tests
Immunology
Primary
Human
Flow Cytometry
25μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Mouse
IgG1, κ
RUO
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal