Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgD Mouse anti-Human, R-PE, Clone: IADB6, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 903009
Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IgD | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Human IgD myeloma protein | |
RUO | |
Human | |
IgG2a |
Flow Cytometry, ELISA | |
IADB6 | |
R-PE | |
Mouse | |
0.1 mg | |
Secondary | |
Store at 2-8°C; Protect from light; Do not freeze |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title