Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Rat anti-Mouse, Brilliant Violet 650, Clone: R35-72, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564208

Catalog No. BDB564208


Add to cart

Description

Description

The rat anti-mouse IgE antibody (clone R35-72) reacts specifically with mouse IgE of the Igh-C [a] and Igh-C [b] haplotypes. It has been reported not to react with other Ig isotypes. Detection with the rat anti-mouse IgE antibody (clone R35-72) of surface immunoglobulin on IgE-secreting hybridoma cells has also been reported.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

IgE
R35-72
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Igh-7; immunoglobulin heavy chain 7 (heavy chain of IgE)
Affinity Purified
Mouse IgE (pooled)
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit