Learn More
MilliporeSigma™ anti-IGF-I Alexa Fluor 488, Clone: Sm1.2,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 05172AF488
Description
Specifically detects IGF-I Clone: Sm1.2 in Chicken, Fish, Human, Mouse, Rat samples, and it is validated for Immunocytochemistry
Insulin-like growth factor I (UniProt P05019; also known as IGF-I, Somatomedin-C) is encoded by the IGF1 (also known as IGFI, MGF) gene (Gene ID 3479) in human. Human insulin-like growth factor-I (IGF-I) is a single chain 70 amino acid polypeptide that shares structural homology with insulin and human IGF-II. IGF-I plays a critical role in growth, differentiation and development via autocrine, paracrine and endocrine mechanisms. It is a mitogen and also has effects on cell migration and differentiation. IGF-I is a regulator of metabolism and is also implicated in the pathophysiology of cancer, diabetes and obesity. Upon binding the tetrameric (alpha:beta)2 IGF-I receptor, the receptor alpha chains induce the tyrosine autophosphorylation of the beta chains, which in turn trigger cell type-specific downstream signaling cascade of events.Specifications
IGF-I | |
Monoclonal | |
Alexa Fluor 488 | |
P05019 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
IGF-I (Russel, W.E., (1984). Proc. Natl. Acad. Sci. U. S. A. 81(8):2389-2392). 40% cross reactivity to IGF-II (Van Wyk, J.J., and Bruton, E.T. (1997). Endocrinology. 138(10):4521-4522). | |
2°C to 8°C for one year from date of shipment | |
IgG1 κ |
Immunocytochemistry | |
Sm1.2 | |
Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide. | |
IGF1, IBP1 | |
Highly purified human plasma insulin-like growth factor I (IGF-I; Somatomedin C; Sm-C) conjugated to mouse albumin (Van Wyk, J.J., and Bruton, E.T. (1997). Endocrinology. 138(10):4521-4522). Action of somatomedins on cell growth: Effect of selective neutralisation of somatomedin C (IGF-I) with a Monoclonal. In: Raiti, S., Tolman, and R.A. (eds.), Human Growth Hormone. pp. 585–599. Plenum Publishing Corp., New York, NY). | |
100 μL | |
Signaling | |
NP_000609 | |
Chicken, Fish, Human, Mouse, Rat | |
Purified |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only