Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-II Mouse anti-Human, Clone: 75015, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen IGF-II
Immunogen E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1
Clone 75015
Applications Neutralization
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB292SP
View Documents
R&D Systems
MAB292SP
25 ug
Each for $125.00
Add to cart
 
MAB292100
View Documents
R&D Systems
MAB292100
100 ug
Each for $377.00
Add to cart
 
MAB292500
View Documents
R&D Systems
MAB292500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IGF-II Monoclonal antibody MAB292 Specifically detects IGF-II in Human samples. It is validated for Neutralization
Specifications

Specifications

IGF-II
75015
Unconjugated
P01344.1
Monoclonal
Primary
Detects human IGF-II in direct ELISA and Western blots. This antibody shows 100% cross-reactivity with recombinant mouse (rm) IGF-II and no cross-reactivity with recombinant human IGF-I or rmIGF-I.
Protein A or G purified from ascites
E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1
Neutralization
Purified
Mouse
IgG1
3481
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit