Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IGF-II R/Mannose 6 Phosphate Receptor (Cation independent), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131957

Catalog No. NBP131957


Add to cart

Description

Description

IGF-II R/IGF2R Polyclonal antibody specifically detects IGF-II R/IGF2R in Human samples. It is validated for Western Blot.
Specifications

Specifications

IGF-II R/Mannose 6 Phosphate Receptor (Cation independent)
0.58mg/mL
Western Blot 1:100-1:2000
Affinity Purified
IGF2R
Protein antigen contains a sequence within a region between 31 and 210 of the human IGF2R protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
300 kDa mannose 6-phosphate receptor, cation-independent mannose-6 phosphate receptor, cation-independent mannose-6-phosphate receptor, CD222, CD222 antigen, CI Man-6-P receptor, CIMPR, CI-MPR, IGF-II receptor, insulin-like growth factor 2 receptorM6P/IGF2R, Insulin-like growth factor II receptor, M6P/IGF2 receptor, M6PR, M6P-R, MPR 300, MPR1, MPRIM6PR
Rabbit
IgG
0.1 ml
Cancer, Golgi Apparatus Markers, Immunology, Innate Immunity
Primary
3482
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only