Learn More
IGF1R Mouse anti-Human, Clone: 3B7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110853
Catalog No. 01-671-866
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10853 detects IGF-1 Receptor from human samples. MA1-10853 has been successfully used in immunoprecipitation applications. The MA1-10853 immunogen is purified human placental IGF-1 receptors.
The insulin-like growth factors, isolated from plasma, are structurally and functionally related to insulin but have a much higher growth-promoting activity. Insulin Like Growth Factor-I is a polypeptide growth factor, which stimulates the proliferation of a wide range of cell types including muscle, bone, and cartilage tissue.
Specifications
Specifications
IGF1R
Monoclonal
1.1 mg/mL
PBS with no preservative; pH 7.4
P08069
CD221, IGFIR, IGFR, JTK13, IGF-I receptor, insulin-like growth factor I receptor, soluble IGF1R variant 1, soluble IGF1R variant 2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3480
Immunoprecipitation
3B7
Unconjugated
IGF1R
Liquid
IGF1R
Purified human placental IGF-1 receptors.
Protein G
RUO
Antibody
Monoclonal
Human