Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF1R alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524610
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
IGF1R alpha Polyclonal antibody specifically detects IGF1R alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
IGF1R alpha | |
Polyclonal | |
Unconjugated | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3480 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P08069 | |
Igf1r | |
KLH conjugated synthetic peptide between 51-77 amino acids from the N-terminal region of human IGF1R | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction