Learn More
IGF2, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA186837
Catalog No. PA186837
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a member of the insulin family of polypeptide growth factors that is involved in development and growth. It is an imprinted gene and is expressed only from the paternally inherited allele. It is a candidate gene for eating disorders. There is a read-through, INS-IGF2, which aligns to this gene at the 3' region and to the upstream INS gene at the 5' region. Alternatively spliced transcript variants, encoding either the same or different isoform, have been found for this gene.
Reconstitute with 0.5 ml PBS containing 0.1% BSA. For long term storage the addition of 0.09% sodium azide is recommended. Prior to reconstitution, store at 4ºC. After reconstitution, store undiluted at -20ºC, avoiding freeze/thaw cycles.This antibody may be used in Western Blotting applications under either reducing or non-reducing conditions.
Specifications
Specifications
IGF2
Polyclonal
Biotin
IGF2
Lyophilized
IGF2
Recombinant human IGF-II
Affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
0.1mg/mL
PBS with no preservative
P01344
C11orf43, FLJ22066, FLJ44734, INSIGF, pp9974
Rabbit
IgG
50μg
4° C
Primary
3481