Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA124773

Catalog No. PA124773

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The insulin-like growth factors, isolated from plasma, are structurally and functionally related to insulin but have a much higher growth-promoting activity.

PA1-24773 detects IGF2 from human samples.PA1-24773 has been successfully used in inhibition assay, and Western blot procedures.The PA1-24773 immunogen is recombinant, full length human protein.
Specifications

Specifications

IGF2
Polyclonal
Unconjugated
IGF2
Purified
IGF2
Recombinant, full length human protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Inhibition Assays, Neutralization, Western Blot
0.25mg/mL
PBS with no preservative
P01344
Insulin Like Growth Factor 2, IGFII
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
3481
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit