Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2 Goat anti-Mouse, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA124774

Catalog No. PA124774


Add to cart

Description

Description

PA1-24774 detects IGF2 from mouse samples. PA1-24774 has been successfully used in neutralizing, ELISA, and Western blot procedures. The PA1-24774 immunogen is recombit, full length mouse protein. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

Specifications

Specifications

IGF2
Polyclonal
Unconjugated
Igf2
Liquid
IGF2
Recombit, full length mouse protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
ELISA, Neutralization, Western Blot
0.1 mg/mL
PBS with 0.1% BSA and no preservative
P09535
Insulin Like Growth Factor 2, IGFII
Goat
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16002
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit