Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523968

Catalog No. PIPA523968


Add to cart

Description

Description

This antibody is predicted to react with chicken, mouse, rat and Xenopus and zebrafish based on sequence homology.

IGF2BP1 is a member of the insulin-like growthfactor 2 mRNA-binding protein family. The protein encoded by thisgene contains four K homology domains and two RNA recognitionmotifs. It functions by binding to the mRNAs of certain genes,including insulin-like growth factor 2, beta-actin andbeta-transducin repeat-containing protein, and regulating theirtranslation. Two transcript variants encoding different isoformshave been found for this gene.
Specifications

Specifications

IGF2BP1
Polyclonal
PBS with 0.09% sodium azide
Q9NZI8
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1, IGF-II mRNA-binding protein 1, IGF2 mRNA-binding protein 1, VICKZ family member 1, ZBP-1, coding region determit-binding protein, insulin-like growth factor 2 mRNA-binding protein 1, zipcode-binding protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10642
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IGF2BP1
Liquid
IGF2BP1
KLH conjugated synthetic peptide between 508-534 amino acids from the C-terminal region of human IGF2BP1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit