Learn More
IGF2BP2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523636
Catalog No. PIPA523636
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a member of the IGF-II mRNA-binding protein (IMP) family. The protein encoded by this gene contains several four KH domains and two RRM domains. It functions by binding to the 5 ft. UTR of the insulin-like growth factor 2 (IGF2) mRNA and regulating IGF2 translation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
IGF2BP2
Polyclonal
Unconjugated
IGF2BP2
Liquid
IGF2BP2
KLH conjugated synthetic peptide between 530-556 amino acids from the C-terminal region of human IGF2BP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9Y6M1
IMP-2, IMP2, VICKZ2, p62, IGF-II mRNA-binding protein 2, IGF2 mRNA-binding protein 2, VICKZ family member 2, hepatocellular carcinoma autoantigen p62, insulin-like growth factor 2 mRNA-binding protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10644