Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-3 Mouse anti-Human, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611504

Catalog No. BDB611504


Add to cart

Description

Description

Insulin-like growth factors (IGF) I and II are peptide hormones that regulate cellular proliferation and differentiation. Most circulating IGFs exist in 130-150kDa ternary complexes containing IGF-Binding Protein-3 (IGFBP-3) and an 85kDa glycoprotein, the acid-labile subunit (ALS). IGFBP-3 is one of six IGFBPs that, by binding IGF peptides, prolong their half life and maintain the IGF reservoir. IGFBP-3 is found in body fluids as multiple 40-50kDa forms as a result of differential glycosylation. The expression of IGFBP-3 in many tissues suggests that it locally modulates the autocrine/paracrine action of IGF peptides. IGFBP-3 binding to fibrinogen may be important for wound healing, since this concentrates IGF-I at wound sites and lowers the affinity of IGF-I for IGFBP-3. In addition, retinoic acid-induced IGFBP-3 expression inhibits the growth promoting effects of IGF-I in breast cancer cells. This may link the retinoid and IGF systems in cell growth regulation and explain how the loss of retinoic acid receptor β leads to breast cancer progression. Thus, IGFBP-3 maintenance and regulation of IGF activity
in various tissues may have diverse physiological roles.

Host Species: Mouse
Clone: 4
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human IGFBP-3 aa. 101-210
Formula Weight [Chemical]: 40 & 44kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

IGFBP-3
4
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
IGF-Binding Protein-3
Affinity Purified
Human IGFBP-3 aa. 101-210
50μg
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit