Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IGFBP-5, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP150718

Catalog No. NBP150718

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IGFBP-5
0.5 mg/mL
Western Blot
NP_000590
IBP5IBP-5, IGF-binding protein 5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5
Rabbit
IgG
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Unconjugated
PBS (pH 7.2) and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
IGFBP5
E. coli expressed human IGF-BP5
Immunogen affinity purified
RUO
Primary
3488
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only