Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525925

Catalog No. PIPA525925


Add to cart

Description

Description

This antibody is predicted to react with bovine and porcine based on sequence homology.

This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
Specifications

Specifications

IGFBP4
Polyclonal
PBS with 0.09% sodium azide
P22692, P47879
BP-4, HT29-IGFBP, IBP4, IGFBP-4, IBP-4, IGF-binding protein 4, insulin-like growth factor-binding protein 4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16010, 3487
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IGFBP4
Liquid
IGFBP4
KLH conjugated synthetic peptide between 96-124 amino acids from the central region of human IGFBP4
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit