Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1; Clone: A85-1; BD

Manufacturer:  BD Biosciences 563285

Catalog No. BDB563285


Add to cart

Description

Description

Host Species: Rat
Clone A85-1
Isotype IgG1, κ
Species Reactivity Ms
Immunogen Pooled Mouse IgG1

Flow Cytometry

Specifications

Specifications

IgG1
0.2mg/mL
Rat
IgG1
RUO
Monoclonal
A85-1
Affinity Purified
Pooled Mouse IgG1
50μg
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit