Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- IgG1 κ, Clone: X40, Mouse, APC-R700, Isotype Control, BD

Isotype Control

Manufacturer:  BD Biosciences 564974

Catalog No. BDB564974


Add to cart

Description

Description

The X40 hybridoma was derived from the fusion of mouse Sp2/0-Ag 14 myeloma cells with spleen cells from BALB/c mice immunized with keyhole limpet hemocyanin (KLH). The hybridoma secretes monoclonal antibody that binds specifically to KLH, an antigen not expressed on human cells or human cell lines.

This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45nm filter).

Applications:

Flow Cytometry, Isotype Control

Specifications

Specifications

Mouse IgG1, kappa
X40
APC-R700
Murine
Mouse (BALB/c) IgG1, κ
4°C, protect from prolonged exposure to light, DO NOT FREEZE
Monoclonal
Flow Cytometry
0.2mg/mL
Affinity Purified
Keyhole limpet hemocyanin (KLH)
RUO
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit