Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, kappa, BB515, Clone: A19-3, BD

Manufacturer:  BD Biosciences 564460

Catalog No. BDB564460


Add to cart

Description

Description

Host Species: Hamster
Clone: A19-3
Isotype: IgG1, κ
Immunogen: TNP-keyhole limpet hemocyanin

Flow Cytometry, Isotype control

Specifications

Specifications

IgG1 κ
BB515
Affinity purified
TNP-keyhole limpet hemocyanin
0.1mg
A19-3
Aqueous buffered solution containing ≤0.09% sodium azide.
Hamster
IgG1, κ
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit