Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG1, kappa, BB515, Clone: X40, BD

Manufacturer:  BD Biosciences 564416

Catalog No. BDB564416


Add to cart

Description

Description

Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Immunogen: Keyhole limpet hemocyanin (KLH)

Flow Cytometry, Isotype control

Specifications

Specifications

IgG1 κ
BB515
Affinity purified
Keyhole limpet hemocyanin (KLH)
100μg
X40
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit