Learn More
anti-IgG1 λ, Rat, PE-Cy™7, Clone: A110-1, Isotype Control, BD
Supplier: BD Biosciences 552869
Description
The A110-1 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, or rat cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone A110-1 was selected as an isotype control following screening for low background on a variety of mouse and rat tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: A110-1
Isotype: IgG1 λ
Immunogen: Trinitrophenal-KLH
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
A110-1 | |
PE-Cyanine7 | |
Trinitrophenol-KLH | |
0.1 mg | |
IgG1 λ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.