Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG2 κ, Hamster, V450, Clone: B81-3, Isotype Control, BD
Supplier: BD Biosciences 560563
Description
The B81-3 antibody is specific for keyhole limpet hemocyanin.
Host Species: Hamster
Clone: B81-3
Isotype: IgG2, κ
Immunogen: Keyhole Limpet Hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
0.2mg/mL | |
Hamster | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
B81-3 | |
Violet Fluor 450 (V450) | |
Keyhole limpet hemocyanin (KLH) | |
0.1 mg | |
IgG2 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction