Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- IgG2a, κ, Clone: R35-95, Rat, APC-R700, Isotype Control, BD

Isotype Control

Manufacturer:  BD Biosciences 564982

Catalog No. BDB564982


Add to cart

Description

Description

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.

This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45nm filter).

Applications:

Flow Cytometry, Isotype Control

Specifications

Specifications

Rat IgG2a, kappa
R35-95
APC-R700
Rat
Rat (LOU) IgG2a, κ
4°C, protect from prolonged exposure to light, DO NOT FREEZE
Monoclonal
Flow Cytometry
0.2mg/mL
Affinity Purified
Mouse Pooled Immunoglobulin
RUO
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit