Learn More
anti-IgG2a κ, Mouse, Alexa Fluor™ 647, Clone: G155-178, BD
Supplier: BD Biosciences 557715
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
Alexa Fluor 647 | |
TNP-keyhole limpet hemocyanin | |
100 Tests | |
IgG2a κ |
G155-178 | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.