Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a κ, Rat, APC-H7, Clone: R35-95, Isotype Control, BD

Manufacturer:  BD Biosciences 560197

Catalog No. BDB560197


Add to cart

Description

Description

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues.

Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Mouse Pooled Immunoglobulin

Flow Cytometry, Isotype Control

Specifications

Specifications

Flow Cytometry
0.2mg/mL
Affinity Purified
Mouse Pooled Immunoglobulin
0.1mg
Murine
R35-95
APC-H7
Rat
IgG2a, k
Undiluted at 4°C
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit