Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2b κ, Mouse, PE-Cy™7, Clone: 27-35, Isotype Control, BD

Manufacturer:  BD Biosciences 560542

Catalog No. BDB560542


Add to cart

Description

Description

The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was s

Host Species: Mouse
Clone: 27-35
Isotype: IgG2b, κ

Flow Cytometry, Isotype Control

Specifications

Specifications

Flow Cytometry
0.2mg/mL
Affinity Purified
IgG2b, k
Undiluted at 4°C. Protect from exposure to light. Do not freeze.
27-35
PE-Cyanine7
Mouse
0.1mg
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit