Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgM, Brilliant Violet 650, Clone: R6-60.2, BD

Manufacturer:  BD Biosciences 564027

Catalog No. BDB564027


Add to cart

Description

Description

Host Species: Rat
Clone: R6-60.2
Isotype: IgG2a, κ
Species Reactivity: Ms
Immunogen: Pooled Mouse Ig

Flow Cytometry

Specifications

Specifications

IgM
0.2mg/mL
Affinity Purified
IgG2a, κ
Secondary
Murine
R6-60.2
Brilliant Violet 650
Rat
50μg
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit