Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgM, kappa, BV650, Clone: G155-228, BD
Supplier: BD Biosciences 564367
Description
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Immunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Isotype control
Specifications
IgM, kappa | |
Brilliant Violet 650 | |
TNP-Keyhole Limpet Hemocyanin | |
50 μg |
G155-228 | |
Mouse | |
Affinity Purified | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction