Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgM, lambda, BV421 , Clone: G235-1, BD

Manufacturer:  BD Biosciences 564340

Catalog No. BDB564340


Add to cart

Description

Description

Host Species: Hamster
Clone: G235-1
Isotype: IgM, lambda1
Immunogen: TNP-keyhole limpet hemocyanin

Flow Cytometry, Isotype control

Specifications

Specifications

IgM, lambda
Brilliant Violet 421
Affinity purified
TNP-keyhole limpet hemocyanin
50μg
G235-1
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Hamster
IgM, lambda1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit