Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524225

Catalog No. PIPA524225


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

This gene encodes a member of the immunoglobulin-like domain-containing superfamily. Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

IGSF1
Polyclonal
PBS with 0.09% sodium azide
Q8N6C5, Q7TQA1
IGCD1, IGDC1, INHBP, PGSF2, p120, immunoglobulin superfamily member 1, immunoglobulin-like domain-containing protein 1, inhibin-binding protein, pituitary gland-specific factor 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
209268, 3547
Immunohistochemistry, Western Blot
Unconjugated
IGSF1
Liquid
IGSF1
KLH conjugated synthetic peptide between 1270-1297 amino acids from the C-terminal region of human IGSF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit