Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IHPK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132322

Catalog No. NBP132322


Add to cart

Description

Description

IHPK2 Polyclonal antibody specifically detects IHPK2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IHPK2
0.51mg/mL
Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
IP6K2
Recombinant protein encompassing a sequence within the center region of human IP6K2. The exact sequence is proprietary.
49 kDa
0.1 ml
Apoptosis
Primary
51447
Human
Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EC 2.7.4.21, IHPK2ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, inositol hexakisphosphate kinase 2, inositol hexaphosphate kinase 2, InsP6 kinase 2, P(i)-uptake stimulator, pi uptake stimulator, PIUS
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Chicken (87%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only