Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-β III Tubulin, (Similar to TuJ1 antigen) ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB9354/DEL

Description
Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications
Β III Tubulin, (Similar to TuJ1 antigen) | |
Unconjugated | |
Synthetic peptides from human/rat β III Tubulin. | |
500 μL | |
Neuroscience | |
Human, Mouse, Rat |
Polyclonal | |
Chicken | |
Affinity Purified | |
RUO | |
Primary | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction