Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKAP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14163

Catalog No. PA14163


Add to cart

Description

Description

PA1-4163 detects IKAP in human samples. PA1-4163 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing IKAP in Interleukin-1-stimulated 293 cells. The PA1-4163 immunogen is a synthetic peptide corresponding to residues I(148) H Q D D F G E S K F I T V(161) of huamn IKAP. This sequence is 92% conserved in rat, and 85% conserved in mouse.

The transcription factor NF-kappaB coordinates the activation of numerous genes in response to pathogens and pro-inflammatory cytokines, and is, therefore, vital in the development of acute and chronic inflammatory diseases. NF-kappaB is activated by cytokine-activated IKB kinases (IKKs); IKK-alpha and IKK-beta isozymes are found in large complexes These large, interleukin-1-inducible IKK complexes have been found to contain a new protein, termed IKK-complex-associated protein (IKAP), which can bind NIK and IKKs and assemble them into an active kinase complex. IKAP is a scaffold protein and a regulator for three different kinases involved in pro-inflammatory cytokine signaling.  
Specifications

Specifications

IKAP
Polyclonal
Unconjugated
ELP1
Liquid
ELP1
Synthetic peptide corresponding to residues I(148) H Q D D F G E S K F I T V(161) of huamn IKAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O95163
IkappaB kinase complex-associated protein, IKK complex-associated protein, p150
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
8518
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit