Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ikaros Mouse anti-Human, Alexa Fluor 488, Clone: R32-1149, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564867

Catalog No. BDB564867


Add to cart

Description

Description

The R32-1149 monoclonal antibody specifically binds to Ikaros, which is encoded by IKZF1 (IKAROS family zinc finger 1). Ikaros is also known as Lymphoid transcription factor LyF-1 (LYF1) and ZNFN1A1 [Zinc finger protein, subfamily 1A, 1 (Ikaros)]. Ikaros belongs to the IKAROS family of zinc-finger transcription factors. This DNA-binding transcription factor forms homodimers and heterodimers with other IKAROS family members. It is involved in the development of lymphoid tissues and remains abundantly expressed in the thymus and by cells within peripheral lymphoid tissues. Ikaros functions in the maturation, differentiation, and homeostasis of various hematopoietic cells including T lymphocytes, B cells, NK cells, and neutrophils. Ikaros regulates the expression of a number of genes including various genes expressed during early stages of B- and T-cell development. Several alternatively-spliced human Ikaros isoforms have been described. Abnormal expression of different Ikaros isoforms has been associated with hematologic malignancies . Flow cytometric analysis of 293F cells transfected with members of the IKAROS family has demonstrated that clone R32-1149 is negative on Helios, Aiolos, and Eos transfectants.

Specifications

Specifications

Ikaros
R32-1149
IKZF1; LYF1; hIk-1; IK1; ZNFN1A1
Affinity purified
Human Ikaros Recombinant Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 488
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit