Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IkB-alpha, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $261.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Within the range of amino acids 130-164 of human IkB alpha protein were used as the immunogen.
Antigen IkB-alpha
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP223901SS
View Documents
Novus Biologicals
NBP223901SS
0.025 ml
Each for $119.60
Add to cart
 
NBP223901
View Documents
Novus Biologicals
NBP223901
0.1 ml
Each for $261.50
Add to cart
 
Description

Description

IkB-alpha Polyclonal antibody specifically detects IkB-alpha in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Rabbit
Within the range of amino acids 130-164 of human IkB alpha protein were used as the immunogen.
Polyclonal
Primary
Western Blot, ELISA
4792
IgG
IkB-alpha
Antisera
RUO
P25963
Crude antibody whole antisera, 50% Glycerol with 0.025% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit