Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB alpha (Cleaved) Mouse anti-Human, Clone: 5D1623, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA141251

Catalog No. 01-676-605


Add to cart

Description

Description

NF-kB is silenced in the cytoplasm by an inhibitory protein, IkB (1). Synthesis of IkBa is autoregulated (2). IkB proteins are phosphorylated by IkB kinase complex consisting of at least three proteins, IKK1/a, IKK2/b, and IKK3/g (3-6). External stimuli such as tumor necrosis factor or other cytokines results in phosphorylation and degradation of IkB releasing NF-kB dimers. NF-kB dimer subsequently translocates to the nucleus and activates target genes. Six members of IkB family members have been identified (1). Recent studies have shown that IkBa can be cleaved by caspases during apoptosis to produce an amino-terminal truncated IkBa (DN-IkBa) (7,8). The DN-IkBa is resistant to degradation in response to inducers of NF-kB and act as dominant inhibitory molecule that suppresses NF-kB activity during apoptosis.

Suggested positive control: anti-Fas treated Jurkat (30 min).
Specifications

Specifications

IkB alpha (Cleaved)
Monoclonal
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P25963
IKBA, MAD-3, NFKBI
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4792
Western Blot
5D1623
Unconjugated
NFKBIA
Liquid
NFKBIA
Caspase-3 mediated cleavage site of IkBa.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit