Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IKK beta, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP176935

Catalog No. NBP176935

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IKK beta
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
O14920
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This polyclonal antibody has no cross response to IKKa or IKKy.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
IKBKB
Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human IKK beta (Genbank accession NoO14920), which differs from corresponding murine sequence by one amino acid. The immunogen is located within amino acids 650 - 700 of IKK beta.
87 kDa
0.1 mg
Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase
Primary
3551
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only