Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKγ Mouse, Unlabeled, Clone: 54, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611306

Catalog No. BDB611306


Add to cart

Description

Description

In most cells, NF-κB is sequestered in an inactive cytoplasmic form via interactions with the inhibitory proteins IκBα, IκBβ, and IκBε. Cell
stimulation induces the release, activation, and nuclear translocation of NF-κB. Release of NF-κB results from the phosphorylation and subsequent proteolytic degradation of the IκB proteins. Two cytokine-inducible IκB kinases (IKKα and IKKβ) phosphorylate and target the IκB proteins for degradation by the ubiquitin pathway. These kinases are components of a 700-900kDa multisubunit complex that also contains NF-κB/RelA, IκBα, MEKK1, NIK, IKAP, and IKKγ/NEMO (NF-κB essential modulator). IKKγ contains two coiled-coil domains and a leucine zipper which allow it to form dimers and trimers that interact directly with IKKβ. IKKγ, essential for IKKα/IKKβ activation of NF-κB, is located functionally and physically upstream of these subunits. In addition, IKKγ enables the HTLV-1 Tax oncoprotein to interact with IKKβ, resulting in constitutive activation of NF-κB and maintenance of cellular transformation. Thus, IKKγ is an essential element of the IκB complex and an adaptor that mediates stable formation of Tax-IKK complexes.

Host Species: Mouse
Clone: 54
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse IKKγ/NEMO aa. 278-396
Formula Weight [Chemical]: 48kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

IKKγ
54
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
NEMO
Affinity Purified
Mouse IKKγ/NEMO aa. 278-396
50μg
Cell Biology
Primary
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit