Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14162

Catalog No. PA14162


Add to cart

Description

Description

PA1-4162 detects IKK gamma in human samples. This antibody recognizes both human IKK gamma isoforms. PA1-4162 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~50 kDa protein representing the IKK gamma protein in Interleukin-1-stimulated 293 cells. The PA1-4162 immunogen is a synthetic peptide corresponding to residues R(385) R S P P E E P P D F C C P K(399) of human IKK gamma.

Pro-inflammatory cytokines activate the transcription factor NF-kappaB by stimulating the activity of a protein kinase that phosphorylates IkappaB, an inhibitor of NF-kappaB, at sites that trigger its ubiquitination and degradation. A large, cytokine-responsive IkappaB kinase (IKK) complex contains 2 subunits, IKK-alpha and IKK-beta, which are protein kinases whose function is needed for NF-kappaB activation by pro-inflammatory stimuli. IKK is composed of similar amounts of IKK-alpha, IKK-beta, which are differentially processed forms of a third subunit, IKK-gamma. IKK-gamma interacts preferentially with IKK-beta and is required for the activation of the IKK complex.
Specifications

Specifications

IKK gamma
Polyclonal
Unconjugated
IKBKG
Liquid
IKBKG
Synthetic peptide corresponding to residues R(385) R S P P E E P P D F C C P K(399) of human IKK gamma.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9Y6K9
NF-kappa-B essential modulator, NEMO, NF-kappa-B essential modifier, Inhibitor of nuclear factor kappa-B kinase gamma subunit, IkB kinase gamma subuni
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
8517
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit