Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528606

Catalog No. PIPA528606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes the regulatory subunit of the inhibitor of kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.

PA5-28606 targets IKK gamma in IF, IHC (P), and WB applications and shows reactivity with Human, Mouse, and Rat samples.The PA5-28606 immunogen is recombinant fragment corresponding to a region within amino acids 1 and 226 of Human IKK gamma.
Specifications

Specifications

IKK gamma
Polyclonal
Unconjugated
IKBKG
Purified
IKBKG
Recombinant fragment corresponding to a region within amino acids 1 and 226 of Human IKK gamma
Antigen affinity chromatography
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
8517
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1.03mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9Y6K9, O88522, Q6TMG5
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IP, IP1, IP2, IPD2, NEMO, I-kappa-B kinase subunit gamma, IKKAP1, IKKG, IkB kinase gamma subunit, NF-kappa-B essential modifier, NF-kappa-B essential modulator, NFkappaB essential modulator, ikB kinase subunit gamma, ikB kinase-associated protein 1, incontinentia pigmenti, inhibitor of nucl
Rabbit
IgG
100μL
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit