Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 alpha/IL-1F1 Membrane Form Mouse anti-Human, Clone: 3405, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen IL-1 alpha/IL-1F1
Immunogen E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583
Clone 3405
Applications Flow Cytometry, Neutralization, CyTOF
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2001SP
View Documents
R&D Systems
MAB2001SP
25 ug
Each for $119.60
Add to cart
 
MAB2001100
View Documents
R&D Systems
MAB2001100
100 ug
Each for $377.00
Add to cart
 
MAB2001500
View Documents
R&D Systems
MAB2001500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-1 alpha/IL-1F1 Membrane Form Monoclonal antibody MAB2001 Specifically detects IL-1 alpha/IL-1F1 Membrane Form in Human samples. It is validated for Flow Cytometry, Neutralization
Specifications

Specifications

IL-1 alpha/IL-1F1
3405
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
3552
RUO
E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583
Flow Cytometry, Neutralization, CyTOF
Purified
P01583
Monoclonal
Primary
Detects human IL-1 alpha/IL-1F1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-1 alpha , recombinant rat IL-1 alpha , recombinant cotton rat IL-1 alpha , recombinant human (rh) IL-1 beta or rhIL-18 is observed.
Protein A or G purified from ascites
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit