Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 beta/IL-1F2 Mouse anti-Porcine, Clone: 77724, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen IL-1 beta/IL-1F2
Immunogen E. coli-derived recombinant porcine IL-1 beta/IL-1F2 Ala115-Pro267 Accession # P26889.
Clone 77724
Applications Western Blot, Neutralization, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB6811SP
View Documents
R&D Systems
MAB6811SP
25 ug
Each for $114.50
Add to cart
 
MAB6811100
View Documents
R&D Systems
MAB6811100
100 ug
Each for $377.00
Add to cart
 
MAB6811500
View Documents
R&D Systems
MAB6811500
500 ug
Each for $839.00
Add to cart
 
Description

Description

IL-1 beta/IL-1F2 Monoclonal antibody MAB6811 Specifically detects IL-1 beta/IL-1F2 in Porcine samples. It is validated for ELISA Capture, Neutralization, Western Blotting
Specifications

Specifications

IL-1 beta/IL-1F2
77724
Unconjugated
P26889.
Monoclonal
Primary
Detects porcine IL-1 beta in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) IL-1 beta and recombinant mouse (rm) IL-1 beta is observed and no cross-reactivity with rhIL-1 alpha , rmIL-1 alpha , recombinant porcine IL-1 alpha or recombinant rat IL-1 alpha is observed.
Protein A or G purified from hybridoma culture supernatant
E. coli-derived recombinant porcine IL-1 beta/IL-1F2 Ala115-Pro267 Accession # P26889.
Western Blot, Neutralization, ELISA
Purified
Mouse
IgG1
3553
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit