Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1RAcP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130896

Catalog No. PIPA130896


Add to cart

Description

Description

PA1-30896 detects IL1 RaCP from human samples. PA1-30896 has been successfully used in Western blot applications. The PA1-30896 immunogen is: Synthetic peptide corresponding to amino acids 549-569 of human IL-1 RAcP

Specifications

Specifications

IL-1RAcP
Polyclonal
Unconjugated
IL1RAP
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3556
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NPH3
IL1RAP
Synthetic peptide corresponding to amino acids 549-569 of human IL-1 RAcP
IgG fraction
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit