Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL1R2 Mouse anti-Human, PE, Clone: 34141, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523543

Catalog No. PIMA523543


Add to cart

Description

Description

When used in combination with the biotinylated human IL-1 RII affinity purified polyclonal detection antibody in sandwich ELISAs, no significant cross-reactivity or interference was observed with recombit human (rh) IL-1ra, rhIL-1 RI, recombit mouse IL-1ra, or recombit rat IL-1ra.

The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.
Specifications

Specifications

IL1R2
Monoclonal
PE
IL1R2
Liquid
IL1R2
Sf 21-derived recombit human IL-1 RII Phe14-Glu343 (Ser56Gly and Glu297Gly)
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
34141
Buffered saline solution with BSA and 0.09% sodium azide
P27930
CD121b, IL1RB, CD121 antigen-like family member B, CDw121b, IL-1 type II receptor, IL-1R-2, IL-1R-beta, IL-1RT-2, IL-1RT2, antigen CDw121b, interleukin-1 receptor beta, interleukin-1 receptor type 2, interleukin-1 receptor type II, type II interleukin-1 receptor, beta, ILR2
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
7850
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit