Learn More
anti-IL-1 RI, PE, Clone: 129304, R&D Systems™
Manufacturer:  R&D Systems FAB7712P025
 View more versions of this product
Catalog No. FAB7712P025
Add to cart
Specifications
Specifications
IL-1 RI
129304
Flow Cytometry 10 uL/10^6 cells
P13504
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Rat
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse IL-1 RI in direct ELISAs and Western blots.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
IL1R1
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI, Leu20-Lys338, Accession # P13504
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3554
Mouse