Learn More
anti-IL-1 RI, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130929
Catalog No. NBP130929
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
IL-1 RI
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
IL1R1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human IL1 Receptor 1. The exact sequence is proprietary.
65 kDa
100 ul
Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3554
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.