Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-1 RII, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP152171

Catalog No. NBP152171

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

IL-1 RII
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
IL1R2
Peptide with sequence C-QYNITRSIELRIKK corresponding to internal region according to NP_004624.1, NP_001248348.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to cross-react with both reported isoforms (NP_004624.1; NP_001248348.1). Reported variants represent identical protein: NP_775465.1, NP_004624.1
ELISA
Peptide ELISA Detection limit 1:2000
NP_004624.1
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Goat
IgG
0.1 mg
Cancer, Signal Transduction
Primary
7850
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only