Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-1 RII, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

Manufacturer:  Novus Biologicals NBP132681

Catalog No. NBP132681


Add to cart

Description

Description

IL-1 RII Polyclonal specifically detects IL-1 RII in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

IL-1 RII
1.0mg/mL
Western Blot 1:500-1:3000, Chromatin Immunoprecipitation, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:500
Affinity Purified
IL1R2
Recombinant protein encompassing a sequence within the center region of human IL1 Receptor 2. The exact sequence is proprietary.
45 kDa
100 ul
Cancer, Signal Transduction
Primary
7850
Human
Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Expected to cross react based on sequence identity: Monkey (94%).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only