Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-12 p70 Mouse anti-Human, Clone: 24945, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523715

Catalog No. PIMA523715


Add to cart

Description

Description

In sandwich ELISAs, no cross-reactivity with recombit human (rh) IL-12 p40, rhIL-12 p35, rhIL-23 or recombit mouse IL-12 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Interleukin-12 (IL-12) is a heterodimeric 70 kDa cytokine composed of two covalently linked, glycosylated chains with molecular weights of 40kD (p40) and 35-kD (p35). IL-12 is mainly produced by monocytes, macrophages, and dendritic cells in response to bacterial products such as lipopolysaccharides (LPS), to intracellular pathogens or upon interaction with activated T cells. IL-12 was originally discovered because of its ability to induce interferon-gamma (IFN-g) production, cell proliferation, and cytotoxicity mediated by natural killer cells and T cells. Studies have established that IL-12 also plays a key role in the development of Th1 responses, leading to IFN-g and IL-2 production. These cytokines can in turn promote T-cell responses and macrophage activation. Recombit mouse IL-12 p70 is produced in baculovirus-infected insect cells as an authentic heterodimer of precursor p35 and p40 subunits using a dual promoter expression system. IL-12 p70 is distinct from other available forms of the protein in that it is expressed as a true heterodimer, as opposed to a single-chain, pseudo-heterodimer in which the subunits are joined by an artificial linker. The responses of lymphocytes to this cytokine are mediated by the activator of transcription protein STAT4. Nitric oxide synthase 2A (NOS2A/NOS2) is found to be required for the signaling process of this cytokine in innate immunity. Human and mouse IL-12 p40 share about 70% amino acid sequence homology.
Specifications

Specifications

IL-12 p70
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P29459, P29460
IL12A, IL12B, Interleukin-12 subunit alpha, IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35, NK cell stimulatory factor chain 1, NKSF1, NKSF1, Interleukin-12 subunit beta, IL-12B, Cytotoxic lymphocyte maturation factor 40 kDa subunit, IL-12 subunit p40, NK cell stimulatory factor chain 2, NKSF2, NKSF2, IL12p35, IL12P40
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3592, 3593
ELISA, Immunocytochemistry, Immunofluorescence
24945
Unconjugated
IL12B
Lyophilized
IL12A, IL12B
Sf 21-derived recombit human IL-12 heterodimer Arg23-Ser219 of p35; Ile23-Ser328 of p40
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit